Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride dihydrate ≥99%

Supplier: BeanTown Chemical
Synonyms: Calcium Chloride Dihydrate

135710-500G 135710-2KG
BT135710-2KGEA 138.51 USD InStock
BT135710-2KG BT135710-500G
Calcium chloride dihydrate ≥99%
Calcium chloride dihydrate
  • Hygroscopic
  • Granular


Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Melting Pt: 176 °C (dec.)
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty