Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium ≥99.9% (trace metals basis), granules -15 mesh

Supplier: BeanTown Chemical

136045-250G 136045-50G 136045-10G
BT136045-250GEA 541.47 USD InStock
BT136045-250G BT136045-50G BT136045-10G
Titanium ≥99.9% (trace metals basis), granules -15 mesh
Titanium
  • RTECS: XR1700000
  • Solid


Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3287 °C
Melting Pt: 1660 °C
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty