Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraammineplatinum(II) chloride monohydrate

Supplier: BeanTown Chemical
Synonyms: Tetraammineplatinum (II) chloride hydrate

136410-5G 136410-1G
BT136410-5GEA 717.74 USD InStock
BT136410-5G BT136410-1G
Tetraammineplatinum(II) chloride monohydrate
Tetraammineplatinum(II) chloride monohydrate
  • Hygroscopic
  • Crystalline


Formula: Cl₂H₁₄N₄OPt
MW: 352.123 g/mol
Melting Pt: 250 °C (dec.)
Density: 2.74 g/mL
MDL Number: MFCD00149947
CAS Number: 13933-33-0
EINECS: 237-706-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty