Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide 20% (w/w) in water, colloidal dispersion

Supplier: BeanTown Chemical
Synonyms: Aluminium(III) oxideAlumina C

136990-1KG
BT136990-1KGEA 48.31 USD InStock
BT136990-1KG
Aluminium oxide 20% (w/w) in water, colloidal dispersion
Aluminium oxide
  • 0.05 Micron Particles


Formula: Al₂O₃
MW: 101.96 g/mol
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty