Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Talc

Supplier: BeanTown Chemical
Synonyms: SoapstoneMagnesium silicate monohydrateTalcum venetum

137650-2.5KG 137650-100G 137650-500G
BT137650-500GEA 42.59 USD InStock
BT137650-500G BT137650-100G BT137650-2.5KG
Talc
Talc
  • RTECS: WW2710000
  • Powder


Formula: Mg₃O₁₁Si₄·1H₂O
MW: 379.27 g/mol
Density: 2.7 g/mL
MDL Number: MFCD00792903
CAS Number: 14807-96-6
EINECS: 238-877-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty