Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Tartaric acid ≥99%

Supplier: BeanTown Chemical
Synonyms: Racemic Acid

137840-1KG 137840-250G
BT137840-1KGEA 142.28 USD InStock
BT137840-1KG BT137840-250G
DL-Tartaric acid ≥99%
DL-Tartaric acid
  • Solid


Formula: C₄H₆O₆
MW: 150.09 g/mol
Melting Pt: 210…212 °C (dec.)
MDL Number: MFCD00071626
CAS Number: 133-37-9
EINECS: 205-105-7
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty