Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(IV) oxide, NanoArc®

Supplier: BeanTown Chemical
Synonyms: Tin dioxideStannic OxideTin(iv)oxide

139275-25G 139275-100G 139275-500G
BT139275-100GEA 106.18 USD InStock
BT139275-100G BT139275-25G BT139275-500G
Tin(IV) oxide, NanoArc®
Tin (IV) oxide
  • RTECS: XQ4000000
  • Refractive index: 1.997
  • 13-19nm APS Powder, S.A. 45-65 m²/g


Formula: O₂Sn
MW: 150.71 g/mol
Boiling Pt: 1800…1900 °C
Melting Pt: 1630 °C
Density: 6.95 g/mL
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty