Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Quinolinol ≥99%

Supplier: BeanTown Chemical
Synonyms: 8-Hydroxyquinoline

139295-100G 139295-2.5KG 139295-500G
BT139295-2.5KGEA 460.13 USD InStock
BT139295-2.5KG BT139295-100G BT139295-500G
8-Quinolinol ≥99%
8-Quinolinol
  • RTECS: VC4200000
  • Light Sensitive
  • Crystalline


Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 267 °C/752 mmHg
Melting Pt: 70…73 °C
MDL Number: MFCD00006807
CAS Number: 148-24-3
EINECS: 205-711-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/52/770552.jpg
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty