Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese ≥99.9% (trace metals basis), irregular pieces

Supplier: BeanTown Chemical

139435-250G 139435-1KG 139435-5KG
BT139435-5KGEA 206.67 USD InStock
BT139435-5KG BT139435-250G BT139435-1KG
Manganese ≥99.9% (trace metals basis), irregular pieces
Manganese
  • RTECS: OO9625000
  • Broken Plate


Formula: Mn
Boiling Pt: 1962 °C
Melting Pt: 1244 °C
Density: 7.3 g/mL
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty