Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium dihydrogen orthophosphate ≥97%

Supplier: BeanTown Chemical
Synonyms: Lithium phosphate monobasicLithium dihydrogenorthophosphateLithium dihydrogenphosphateLithium dihydrogen phosphate

139575-250G
BT139575-250GEA 102.22 USD InStock
BT139575-250G
Lithium dihydrogen orthophosphate ≥97%
Lithium dihydrogen orthophosphate
  • Powder


Formula: H₂LiO₄P
MW: 103.93 g/mol
Melting Pt: >100 °C
Density: 2.5 g/mL
MDL Number: MFCD00016188
CAS Number: 13453-80-0
EINECS: 236-633-6
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty