Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl terephthalate ≥99%

Supplier: BeanTown Chemical
Synonyms: Terephthalic acid dimethyl ester

139640-2.5KG 139640-10KG 139640-500G
BT139640-500GEA 37.29 USD InStock
BT139640-500G BT139640-10KG BT139640-2.5KG
Dimethyl terephthalate ≥99%
Dimethyl terephthalate
  • RTECS: WZ1225000
  • Pieces


Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 139…141 °C
MDL Number: MFCD00008440
CAS Number: 120-61-6
EINECS: 204-411-8
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty