Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitro blue tetrazolium chloride (NBT) ≥90%, Technical Grade

Supplier: BeanTown Chemical
Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

140245-500MG
BT140245-500MGEA 93.06 USD InStock
BT140245-500MG
Nitro blue tetrazolium chloride (NBT) ≥90%, Technical Grade
Nitro blue tetrazolium chloride (NBT)
  • RTECS: XF8045000
  • Air Sensitive, Light Sensitive
  • Solid


Formula: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00150013
CAS Number: 298-83-9
EINECS: 206-067-4
Order Now
Specification Test Results
Assay 90%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty