Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-3-hydroxypyridine ≥99%

Supplier: BeanTown Chemical
Synonyms: 2-Bromo-3-pyridinol

140570-5G 140570-100G 140570-25G
BT140570-5GEA 58.07 USD InStock
BT140570-5G BT140570-100G BT140570-25G
2-Bromo-3-hydroxypyridine ≥99%
2-Bromo-3-hydroxypyridine
  • RTECS: UU7705000
  • Powder


Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 185…188 °C
MDL Number: MFCD00006220
CAS Number: 6602-32-0
EINECS: 229-547-5
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty