Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol purple sodium salt 0.04% (w/v) in water

Supplier: BeanTown Chemical
Synonyms: Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolateSodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonateSodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulfonateBromocresol purple water soluble

141665-500ML 141665-100ML
BT141665-100MLEA 23.9 USD InStock
BT141665-100ML BT141665-500ML
Bromocresol purple sodium salt 0.04% (w/v) in water
Bromocresol purple sodium salt
  • Liquid


Formula: C₂₁H₁₅Br₂NaO₅S
MW: 562.21 g/mol
Density: 0.991 g/mL
MDL Number: MFCD00148896
CAS Number: 62625-30-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty