Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyruvaldehyde (methylglyoxal) 35 - 45% (w/w) in water

Supplier: BeanTown Chemical

142150-100G 142150-25G 142150-500G
BT142150-500GEA 210.85 USD InStock
BT142150-500G BT142150-25G BT142150-100G
Pyruvaldehyde (methylglyoxal) 35 - 45% (w/w) in water
Pyruvaldehyde (Methylglyoxal)
  • Refractive index: 1.405
  • Air Sensitive
  • Liquid


Formula: C₃H₄O₂
Density: 1.17 g/mL
MDL Number: MFCD00006960
CAS Number: 78-98-8
EINECS: 201-164-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty