Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-4-bromobutyrate ≥98%

Supplier: BeanTown Chemical
Synonyms: Ethyl 4-bromobutyrate

142200-1KG 142200-250G 142200-50G
BT142200-250GEA 185.67 USD InStock
BT142200-250G BT142200-1KG BT142200-50G
Ethyl-4-bromobutyrate ≥98%
Ethyl 4-bromobutyrate
  • Refractive index: 1.456
  • Liquid


Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Boiling Pt: 80…82 °C/10 mmHg
Density: 1.363 g/mL
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00000259
CAS Number: 2969-81-5
EINECS: 221-005-6
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty