Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexaboron silicide ≥98% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Boron silicide

142295-10G 142295-50G
BT142295-50GEA 311.6 USD InStock
BT142295-50G BT142295-10G
Hexaboron silicide ≥98% (trace metals basis)
Hexaboron silicide
  • -200 Mesh Powder


Formula: B₆Si
MDL Number: MFCD00180907
CAS Number: 12008-29-6
EINECS: 234-535-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty