Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium(II) oxide monohydrate ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Palladium(II) oxide hydratePalladium monoxide monohydrate

142720-1G 142720-5G
BT142720-1GEA 112 USD InStock
BT142720-1G BT142720-5G
Palladium(II) oxide monohydrate ≥99.9% (trace metals basis)
Palladium (II) oxide monohydrate
  • Powder


Formula: OPd·1H₂O
MW: 140.43 g/mol
MDL Number: MFCD00011172
CAS Number: 64109-12-2
EINECS: 215-218-3
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty