Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA tripotassium salt

Supplier: BeanTown Chemical
Synonyms: Edetic acid tripotassium saltTripotassium hydrogen ethylenediaminetetraacetatePotassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetateEdta-3kEdetate TripotassiumEthylenedinitrilotetra-acetic acid tripotassium saltEthylenediaminetetra-acetic acid tripotassium salt

143170-100G 143170-25G
BT143170-100GEA 114.25 USD InStock
BT143170-100G BT143170-25G
EDTA tripotassium salt
EDTA tripotassium salt
  • Crystalline


Formula: C₁₀H₁₃K₃N₂O₈
MW: 406.52 g/mol
MDL Number: MFCD00013304
CAS Number: 17572-97-3
EINECS: 241-543-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty