Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(II) 2-ethylhexanoate, Technical Grade

Supplier: BeanTown Chemical

144265-100G 144265-500G 144265-2KG
BT144265-500GEA 86.42 USD InStock
BT144265-500G BT144265-2KG BT144265-100G
Tin(II) 2-ethylhexanoate, Technical Grade
Tin (II) 2-ethylhexanoate
  • RTECS: MO7870000
  • Refractive index: 1.494
  • Liquid


Formula: C₁₆H₃₀O₄Sn
MW: 405.12 g/mol
Melting Pt: 9 °C
Density: 1.251 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty