Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical
Synonyms: Aluminium(III) oxideAlumina C

144515-25G 144515-100G 144515-500G
BT144515-25GEA 174.95 USD InStock
BT144515-25G BT144515-100G BT144515-500G
Aluminium oxide ≥99.995% (trace metals basis)
Aluminium oxide
  • Powder


Formula: Al₂O₃
MW: 101.96 g/mol
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Order Now
Specification Test Results
Assay 99.995%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty