Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hyodeoxycholic acid ≥96%

Supplier: BeanTown Chemical

145035-5G 145035-100G 145035-25G
BT145035-5GEA 55.47 USD InStock
BT145035-5G BT145035-25G BT145035-100G
Hyodeoxycholic acid ≥96%
Hyodeoxycholic acid
  • RTECS: FZ2050000
  • Solid


Formula: C₂₄H₄₀O₄
Melting Pt: 200…201 °C
MDL Number: MFCD00003681
CAS Number: 83-49-8
EINECS: 201-483-2
Order Now
Specification Test Results
Assay 96%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty