Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid ≥99.5% ACS

Supplier: BeanTown Chemical
Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acidOrthoboric acid

145750-500G 145750-2KG
BT145750-2KGEA 113.21 USD InStock
BT145750-2KG BT145750-500G
Boric acid ≥99.5% ACS
ortho-Boric acid
  • RTECS: ED4550000
  • Granular


Formula: B(OH)₃
MW: 61.83 g/mol
Melting Pt: 160 °C (dec.)
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg
Order Now
Specification Test Results
Assay 99.5% min.
Insoluble in alcohol ≤0.005%
Nonvolatile with methanol ≤0.05%
ph 5% aq. soln 3.6-4.8
Chloride (Cl) ≤0.001%
Phosphate (PO4) ≤0.001%
Arsenic (As) ≤1ppm
Heavy metals ≤0.001%
Sulfate (SO4) ≤0.01%
Calcium (Ca) ≤0.005%
Iron (Fe) ≤0.001%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty