Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(IV) oxide anatase ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

211635-500G 211635-100G 211635-25G
BT211635-500GEA 110.07 USD InStock
BT211635-500G BT211635-25G BT211635-100G
Titanium(IV) oxide anatase ≥99.5% (trace metals basis)
Titanium (IV) oxide anatase
  • RTECS: BV6550000
  • -325 Mesh Powder


Formula: TiO₂
MW: 79.88 g/mol
Melting Pt: 1825 °C
Density: 3.9 g/mL
MDL Number: MFCD00011269
CAS Number: 1317-70-0
EINECS: 215-280-1
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty