Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Povidone iodine

Supplier: BeanTown Chemical
Synonyms: Polyvinylpyrrolidone-iodine complex

211980-1KG 211980-50G 211980-250G
BT211980-250GEA 170.3 USD InStock
BT211980-250G BT211980-50G BT211980-1KG
Povidone iodine
Povidone iodine
  • RTECS: TR1579600
  • Powder


Formula: C₆H₉I₂NO
MW: 364.953 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00084483
CAS Number: 25655-41-8
EINECS: 215-034-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty