Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium ≥99.99% (trace metals basis), foil, Thickness 0.25 mm

Supplier: BeanTown Chemical

212080-100X100 212080-50X50MM 212080-25X25MM
BT212080-25X25EA 99.51 USD InStock
BT212080-25X25 BT212080-100X1 BT212080-50X50
Titanium ≥99.99% (trace metals basis), foil, Thickness 0.25 mm
Titanium
  • RTECS: XR1700000
  • Solid


Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3287 °C
Melting Pt: 1660 °C
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty