Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Inhibitor removal resin

Supplier: BeanTown Chemical

212130-25G 212130-500G 212130-100G
BT212130-25GEA 29.7 USD InStock
BT212130-25G BT212130-100G BT212130-500G
Inhibitor removal resin
Inhibitor removal resin


MDL Number: MFCD00081548
CAS Number: 9003-70-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty