Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl levulinate ≥98%

Supplier: BeanTown Chemical
Synonyms: Levulinic acid ethyl ester4-Oxovaleric acid ethyl ester4-Ketovaleric acid ethyl esterEthyl 4-oxovalerate4-Oxopentanoic acid ethyl esterEthyl 4-oxopentanoate

212365-100G 212365-500G
BT212365-100GEA 43.84 USD InStock
BT212365-100G BT212365-500G
Ethyl levulinate ≥98%
Ethyl levulinate
  • RTECS: OI1700000
  • Refractive index: 1.422
  • Liquid


Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 93…94 °C/18 mmHg
Density: 1.016 g/mL
Flash Pt: 94 °C (201 °F)
MDL Number: MFCD00009209
CAS Number: 539-88-8
EINECS: 208-728-2
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty