Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

penta-Sodium triphosphate

Supplier: BeanTown Chemical
Synonyms: Sodium triphosphate pentabasicSodium tripolyphosphate

212680-2KG 212680-500G
BT212680-2KGEA 85.93 USD InStock
BT212680-2KG BT212680-500G
penta-Sodium triphosphate
Pentasodium triphosphate
  • RTECS: YK4570000
  • Hygroscopic
  • Granular


Formula: Na₅O₁₀P₃
MW: 367.86 g/mol
Melting Pt: 622 °C
MDL Number: MFCD00003514
CAS Number: 7758-29-4
EINECS: 231-838-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty