Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdic acid ≥85.0%

Supplier: BeanTown Chemical

212745-100G 212745-2.5KG 212745-500G
BT212745-100GEA 39.74 USD InStock
BT212745-100G BT212745-2.5KG BT212745-500G
Molybdic acid ≥85.0%
Molybdic acid
  • Crystalline/Powder


Formula: H₂MoO₄
MW: 161.95 g/mol
Melting Pt: 300 °C
Density: 3.1 g/mL
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty