Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium aluminate ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

213465-100G 213465-25G
BT213465-25GEA 70.38 USD InStock
BT213465-25G BT213465-100G
Strontium aluminate ≥99.5% (trace metals basis)
Strontium aluminate
  • -100 Mesh Powder


Formula: Al₂O₄Sr
Density: 3.559 g/mL
MDL Number: MFCD00054054
CAS Number: 12004-37-4
EINECS: 234-455-3
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty