Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Beryllium ≥99.8% (trace metals basis), foil, vacuum tight, Thickness 0.1 mm

Supplier: BeanTown Chemical

213640-50X50MM 213640-25X25MM
BT213640-50X50EA 3091.22 USD InStock
BT213640-50X50 BT213640-25X25
Beryllium ≥99.8% (trace metals basis), foil, vacuum tight, Thickness 0.1 mm
Beryllium
  • Solid


Formula: Be
MW: 9.01 g/mol
Boiling Pt: 2970 °C
Melting Pt: 1278 °C
Density: 1.85 g/mL
MDL Number: MFCD00134032
CAS Number: 7440-41-7
EINECS: 231-150-7
Order Now
Specification Test Results
Assay 99.8%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty