Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis[2-carboxyethylgermanium(IV)] sesquioxide ≥99.5%

Supplier: BeanTown Chemical

215185-25G 215185-5G 215185-100G
BT215185-25GEA 248.22 USD InStock
BT215185-25G BT215185-100G BT215185-5G
Bis[2-carboxyethylgermanium(IV)] sesquioxide ≥99.5%
Bis[2-carboxyethylgermanium(IV)] sesquioxide
  • RTECS: UA2459750
  • Powder


Formula: C₆H₁₀Ge₂O₇
MDL Number: MFCD00049319
CAS Number: 12758-40-6
EINECS: 235-800-0
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty