Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dowex® 1X8 (Cl) 50-100 (Cl), beads

Supplier: BeanTown Chemical

215725-1KG 215725-50G 215725-250G
BT215725-250GEA 206.36 USD InStock
BT215725-250G BT215725-50G BT215725-1KG
Dowex® 1X8 (Cl) 50-100 (Cl), beads
Dowex® 1X8 (Cl)


MDL Number: MFCD00132718
CAS Number: 12627-85-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty