Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium iodide, anhydrous ≥98%

Supplier: BeanTown Chemical

ampouled under argon

215730-250G 215730-50G
BT215730-50GEA 135.69 USD InStock
BT215730-50G BT215730-250G
Lithium iodide, anhydrous ≥98%
Lithium iodide
  • Hygroscopic, Light Sensitive
  • Powder/Lump


Formula: ILi
MW: 133.85 g/mol
Melting Pt: 446 °C
Density: 3.49 g/mL
MDL Number: MFCD00011092
CAS Number: 10377-51-2
EINECS: 233-822-5
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty