Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium ≥99.99% (trace metals basis), rods Ø 3.175 mm

Supplier: BeanTown Chemical

216020-25CM 216020-5CM
BT216020-25CMEA 75.31 USD InStock
BT216020-25CM BT216020-5CM
Titanium ≥99.99% (trace metals basis), rods Ø 3.175 mm
Titanium
  • Solid


Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3287 °C
Melting Pt: 1660 °C
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty