Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(IV) 2-ethylhexanoate ≥97%

Supplier: BeanTown Chemical

216790-500G 216790-100G
BT216790-100GEA 201.46 USD InStock
BT216790-100G BT216790-500G
Titanium(IV) 2-ethylhexanoate ≥97%
Titanium(IV) 2-ethylhexanoate
  • Liquid


Formula: C₃₂H₆₀O₈Ti
MDL Number: MFCD05663781
CAS Number: 3645-34-9
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty