Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) ≥99%

Supplier: BeanTown Chemical
Synonyms: NSC 51986Coniferic acidNSC 2821NSC 6743203-(4-Hydroxy-3-methoxyphenyl)acrylic acidFerulic acid

217130-10G
BT217130-10GEA 33.2 USD InStock
BT217130-10G
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) ≥99%
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
  • RTECS: UD3365500
  • Powder


Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
Melting Pt: 168…172 °C
CAS Number: 1135-24-6
EINECS: 214-490-0
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty