Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-β-Nitrostyrene ≥98%

Supplier: BeanTown Chemical
Synonyms: (E)-(2-Nitrovinyl)benzenetrans-1-Nitro-2-phenylethylene

217940-25G 217940-5G 217940-100G
BT217940-25GEA 98.16 USD InStock
BT217940-25G BT217940-5G BT217940-100G
trans-β-Nitrostyrene ≥98%
trans-β-Nitrostyrene
  • RTECS: WL5460000
  • Crystalline


Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 250…260 °C
Melting Pt: 55…58 °C
MDL Number: MFCD00007402
EINECS: 203-066-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty