Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentyn-1-ol ≥98%

Supplier: BeanTown Chemical
Synonyms: 1-Pentyn-5-ol

218145-25G 218145-100G 218145-5G
BT218145-5GEA 41.45 USD InStock
BT218145-5G BT218145-25G BT218145-100G
4-Pentyn-1-ol ≥98%
4-Pentyn-1-ol
  • Refractive index: 1.445
  • Liquid


Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 154…155 °C
Density: 0.904 g/mL
Flash Pt: 62 °C (144 °F)
MDL Number: MFCD00002974
CAS Number: 5390-04-5
EINECS: 226-383-6
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty