Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl glyoxime ≥99.0% ACS

Supplier: BeanTown Chemical
Synonyms: Diacetyl dioxime2,3-Butanedione dioximeDiacetyl glyoxime

218875-2.5KG 218875-500G 218875-100G
BT218875-500GEA 158.56 USD InStock
BT218875-500G BT218875-100G BT218875-2.5KG
Dimethyl glyoxime ≥99.0% ACS
Dimethyl glyoxime
  • RTECS: EK2975000
  • Solid


Formula: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Melting Pt: 240…241 °C
MDL Number: MFCD00002117
CAS Number: 95-45-4
EINECS: 202-420-1
Order Now
Specification Test Results
Assay 99.0% min.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty