Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzothiazole ≥98%

Supplier: BeanTown Chemical

219675-100G 219675-5G 219675-25G
BT219675-25GEA 114.24 USD InStock
BT219675-25G BT219675-100G BT219675-5G
2-Chlorobenzothiazole ≥98%
2-Chlorobenzothiazole
  • RTECS: DL3150000
  • Refractive index: 1.637
  • Liquid


Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C/30 mmHg
Melting Pt: 21…23 °C
Density: 1.303 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00005776
CAS Number: 615-20-3
EINECS: 210-415-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty