Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl ≥98% free radical

Supplier: BeanTown Chemical
Synonyms: 3-Carboxy-PROXYL3-Carboxy-2,2,5,5-tetramethyl-1-pyrrolidinyloxy

220500-250MG
BT220500-250MGEA 97.12 USD InStock
BT220500-250MG
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl ≥98% free radical
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl
  • Powder


Formula: C₉H₁₆NO₃
MW: 186.23 g/mol
Melting Pt: 200 °C (dec.)
MDL Number: MFCD00003167
CAS Number: 2154-68-9
EINECS: 218-448-2
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty