Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium tetracyanopalladate(II) hydrate

Supplier: BeanTown Chemical

220835-1G
BT220835-1GEA 163.65 USD InStock
BT220835-1G
Potassium tetracyanopalladate(II) hydrate
Potassium tetracyanopalladate(II) hydrate
  • Crystalline


Formula: K₂[Pd(CN)₄]·xH₂O
MW: 288.689 g/mol (anhy)
MDL Number: MFCD00150431
CAS Number: 14516-46-2
EINECS: 238-520-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty