Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium carbonate ≥99.995% (trace metals basis excluding Ca)

Supplier: BeanTown Chemical

221230-50G 221230-1KG 221230-250G
BT221230-50GEA 107.35 USD InStock
BT221230-50G BT221230-250G BT221230-1KG
Lithium carbonate ≥99.995% (trace metals basis excluding Ca)
Lithium carbonate
  • RTECS: OJ5800000
  • Powder


Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 618 °C
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Order Now
Specification Test Results
Assay 99.995%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty