Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rosolic acid (Aurin)

Supplier: BeanTown Chemical
Synonyms: 4-(Bis(4-hydroxyphenyl)methylene)cyclohexa-2,5-dien-1-one

221690-100G 221690-25G
BT221690-25GEA 73.3 USD InStock
BT221690-25G BT221690-100G
Rosolic acid (Aurin)
Rosolic acid (Aurin)
  • Powder


Formula: C₁₉H₁₄O₃
MW: 290.32 g/mol
MDL Number: MFCD00001624
CAS Number: 603-45-2
EINECS: 210-041-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty