Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(II) 2-ethylhexanoate ≥96%

Supplier: BeanTown Chemical

221785-25G 221785-100G
BT221785-25GEA 45.29 USD InStock
BT221785-25G BT221785-100G
Tin(II) 2-ethylhexanoate ≥96%
Tin (II) 2-ethylhexanoate
  • RTECS: MO7870000
  • Refractive index: 1.494
  • Liquid


Formula: C₁₆H₃₀O₄Sn
MW: 405.12 g/mol
Melting Pt: 9 °C
Density: 1.251 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6
Order Now
Specification Test Results
Assay 96%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty