Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Graphite, plate HOPG 10x10x2mm

Supplier: BeanTown Chemical

221875-5PC 221875-1PC
BT221875-1PCEA 576.29 USD InStock
BT221875-1PC BT221875-5PC
Graphite, plate HOPG 10x10x2mm
Graphite
  • Solid, 0.8 ±0.2° mosaic angle


Formula: C
MW: 12.01 g/mol
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty