Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide ACS

Supplier: BeanTown Chemical

221900-10G 221900-50G
BT221900-50GEA 162 USD InStock
BT221900-50G BT221900-10G
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide ACS
Phenylazoformic acid 2-phenylhydrazide compound with 1,5-diphenylcarbohydrazide
  • Solid


Formula: C₂₆H₂₆N₈O₂
Melting Pt: 159…161 °C
MDL Number: MFCD00013218
CAS Number: 10329-15-4
EINECS: 208-698-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty