Synonyms:
L-3-Imidazol-4-ylalanine
, (S)-(+)-Histidine
, (S)-2-Amino-3-(4-imidazolyl)propionic acid
, NSC 137773
, L-Histidine
, H-His-OH
222780-500G
222780-100G
222780-25G
BT222780-25GEA
35.05
USD
InStock
BT222780-25G
BT222780-500G
BT222780-100G
L(+)-Histidine ≥98%
L(+)-Histidine
- RTECS: MS3070000
- Optical rotation: [α]20/D -39±1°, c = 2% in water
- Solid